siRNA Details
Gene Symbol: Wfdc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161806.1 | NP_001155278.1 | 5 | 550 | ||
GenBank mRNA | AK079019.1 | - | 5 | 551 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wfdc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161806.1 | NP_001155278.1 | 5 | 550 | ||
GenBank mRNA | AK079019.1 | - | 5 | 551 |