siRNA Details
| Gene Symbol: Dcpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039238.2 | NP_001034327.2 | 4 | 411 | ||
| GenBank mRNA | BC059096.1 | - | 3 | 430 | 4914089 | |
| DQ237937.1 | - | 411 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcpp2| Gene Symbol: Dcpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039238.2 | NP_001034327.2 | 4 | 411 | ||
| GenBank mRNA | BC059096.1 | - | 3 | 430 | 4914089 | |
| DQ237937.1 | - | 411 | ||||