siRNA Details
| Gene Symbol: Avpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019136.1 | NP_062009.1 | 2 | 820 | ||
| XM_006229549.2 | XP_006229611.1 | 3 | 2214 | |||
| GenBank mRNA | AY338195.1 | - | 2 | 820 | ||
More Information
Other Species Targeted
Set Membership: |
|

Avpr2
Genomic Map