siRNA Details
| Gene Symbol: Gm12250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135115.1 | NP_001128587.1 | 2 | 2595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12250| Gene Symbol: Gm12250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135115.1 | NP_001128587.1 | 2 | 2595 | ||