siRNA Details
| Gene Symbol: Erich4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039243.3 | NP_001034332.2 | 2 | 764 | ||
| GenBank mRNA | AK143993.1 | - | 2 | 721 | ||
| BC038495.1 | - | 2 | 774 | |||
| BC150835.1 | - | 2 | 763 | 9087746 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich4| Gene Symbol: Erich4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039243.3 | NP_001034332.2 | 2 | 764 | ||
| GenBank mRNA | AK143993.1 | - | 2 | 721 | ||
| BC038495.1 | - | 2 | 774 | |||
| BC150835.1 | - | 2 | 763 | 9087746 | ||