siRNA Details
| Gene Symbol: Rergl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128090.1 | NP_001121562.1 | 5 | 596 | ||
| GenBank mRNA | BC147153.1 | - | 5 | 596 | 9056190 | |
| BC147154.1 | - | 5 | 596 | 9056193 | ||
| BC147732.1 | - | 5 | 661 | 9056681 | ||
| BC147735.1 | - | 5 | 671 | 9056684 | ||

Rergl
Genomic Map