siRNA Details
| Gene Symbol: Olfr287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011780.1 | NP_001011780.1 | 3 | 1717 | ||
| GenBank mRNA | AK140728.1 | - | 1 | 1589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr287| Gene Symbol: Olfr287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011780.1 | NP_001011780.1 | 3 | 1717 | ||
| GenBank mRNA | AK140728.1 | - | 1 | 1589 | ||