siRNA Details
| Gene Symbol: Bsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019146.2 | NP_062019.2 | 9 | 11804 | ||
| GenBank mRNA | CB580320.1 | - | 3 | 394 | ||
| Y16563.1 | - | 9 | 11819 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bsn| Gene Symbol: Bsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019146.2 | NP_062019.2 | 9 | 11804 | ||
| GenBank mRNA | CB580320.1 | - | 3 | 394 | ||
| Y16563.1 | - | 9 | 11819 | |||