siRNA Details
| Gene Symbol: Gm9866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044503.1 | - | 2 | 786 | ||
| AK163447.1 | - | 4 | 893 | |||
| BE648300.1 | - | 2 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9866| Gene Symbol: Gm9866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044503.1 | - | 2 | 786 | ||
| AK163447.1 | - | 4 | 893 | |||
| BE648300.1 | - | 2 | 416 | |||