siRNA Details
| Gene Symbol: Cntnap5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077425.1 | NP_001070893.1 | 20 | 3313 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntnap5a| Gene Symbol: Cntnap5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077425.1 | NP_001070893.1 | 20 | 3313 | ||