siRNA Details
Gene Symbol: Strn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019148.2 | NP_062021.1 | 2 | 357 | ||
XM_006239622.2 | XP_006239684.1 | 2 | 453 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Strn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019148.2 | NP_062021.1 | 2 | 357 | ||
XM_006239622.2 | XP_006239684.1 | 2 | 453 |