siRNA Details
| Gene Symbol: Gm7244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101597.2 | NP_001095067.1 | 2 | 205 | ||
| GenBank mRNA | CX238868.1 | - | 2 | 205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7244| Gene Symbol: Gm7244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101597.2 | NP_001095067.1 | 2 | 205 | ||
| GenBank mRNA | CX238868.1 | - | 2 | 205 | ||