siRNA Details
| Gene Symbol: Aadacl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128091.1 | NP_001121563.1 | 2 | 425 | ||
| GenBank mRNA | BC147724.1 | - | 2 | 425 | 9056673 | |
| BC147728.1 | - | 2 | 425 | 9056677 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aadacl2| Gene Symbol: Aadacl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128091.1 | NP_001121563.1 | 2 | 425 | ||
| GenBank mRNA | BC147724.1 | - | 2 | 425 | 9056673 | |
| BC147728.1 | - | 2 | 425 | 9056677 | ||