siRNA Details
| Gene Symbol: Fbln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019153.3 | NP_062026.2 | 6, 7 | 841 | ||
| XM_006240459.2 | XP_006240521.1 | 6, 7 | 1072 | |||
| GenBank mRNA | AF112153.1 | - | 6, 7 | 965 | ||
| AF137350.1 | - | 6, 7 | 768 | |||
| BC078845.1 | - | 6, 7 | 841 | 7123752 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011812, AK085170, AF112151, AK159960, AK159189, and others.

Fbln5
Genomic Map