siRNA Details
| Gene Symbol: Skint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102662.1 | NP_001096132.1 | 8 | 1206 | ||
| GenBank mRNA | EF494889.1 | - | 8 | 1206 | ||
| EF494890.1 | - | 8 | 1206 | |||
| EF494891.1 | - | 8 | 1206 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skint1| Gene Symbol: Skint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102662.1 | NP_001096132.1 | 8 | 1206 | ||
| GenBank mRNA | EF494889.1 | - | 8 | 1206 | ||
| EF494890.1 | - | 8 | 1206 | |||
| EF494891.1 | - | 8 | 1206 | |||