siRNA Details
Gene Symbol: Gm20767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039648.3 | NP_001034737.2 | 1 | 107 | ||
GenBank mRNA | AK139926.1 | - | 1 | 108 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm20767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039648.3 | NP_001034737.2 | 1 | 107 | ||
GenBank mRNA | AK139926.1 | - | 1 | 108 |