siRNA Details
| Gene Symbol: Gm20767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039648.3 | NP_001034737.2 | 1 | 241 | ||
| GenBank mRNA | AK139926.1 | - | 1 | 242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20767| Gene Symbol: Gm20767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039648.3 | NP_001034737.2 | 1 | 241 | ||
| GenBank mRNA | AK139926.1 | - | 1 | 242 | ||