siRNA Details
| Gene Symbol: Dip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081419.2 | NP_001074888.2 | 19 | 2366 | ||
| GenBank mRNA | AB093213.2 | - | 17 | 2036 | ||
| AK019853.1 | - | 14 | 1548 | |||
| AK050040.1 | - | 18 | 2250 | |||
| BC068227.1 | - | 16 | 2005 | |||

Dip2a
Genomic Map