siRNA Details
| Gene Symbol: Dip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081419.2 | NP_001074888.2 | 9 | 1188 | ||
| GenBank mRNA | AB093213.2 | - | 7 | 858 | ||
| AK019853.1 | - | 4 | 370 | |||
| AK050040.1 | - | 8 | 1072 | |||
| AK052899.1 | - | 9 | 1130 | |||
| BC068227.1 | - | 6 | 827 | |||

Dip2a
Genomic Map