siRNA Details
Gene Symbol: Gm7325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177470.1 | NP_001170941.1 | 1 | 89 | ||
GenBank mRNA | CJ062905.1 | - | 1 | 79 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177470.1 | NP_001170941.1 | 1 | 89 | ||
GenBank mRNA | CJ062905.1 | - | 1 | 79 |