siRNA Details
| Gene Symbol: Gm15386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040027.2 | NP_001035116.1 | 2 | 233 | ||
| GenBank mRNA | DQ012028.1 | - | 2 | 175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15386| Gene Symbol: Gm15386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040027.2 | NP_001035116.1 | 2 | 233 | ||
| GenBank mRNA | DQ012028.1 | - | 2 | 175 | ||