siRNA Details
| Gene Symbol: Tctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039153.3 | NP_001034242.2 | 6 | 853 | ||
| GenBank mRNA | AK036568.1 | - | 6 | 835 | ||
| AK138661.1 | - | 6 | 852 | |||
| AK144772.1 | - | 6 | 829 | |||
| DQ278867.1 | - | 6 | 854 | |||

Tctn1
Genomic Map