siRNA Details
| Gene Symbol: Tctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039153.3 | NP_001034242.2 | 11 | 1319 | ||
| GenBank mRNA | AK036568.1 | - | 11 | 1301 | ||
| AK138661.1 | - | 10 | 1241 | |||
| AK144772.1 | - | 11 | 1295 | |||
| DQ278867.1 | - | 11 | 1320 | |||

Tctn1
Genomic Map