siRNA Details
Gene Symbol: Hhla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145096.1 | NP_001138568.1 | 13 | 1209 | ||
GenBank mRNA | AK158329.1 | - | 13 | 1211 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hhla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145096.1 | NP_001138568.1 | 13 | 1209 | ||
GenBank mRNA | AK158329.1 | - | 13 | 1211 |