siRNA Details
| Gene Symbol: Zfp784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039532.2 | NP_001034621.1 | 2 | 1444 | ||
| GenBank mRNA | AK050452.1 | - | 2 | 1439 | ||
| AK161791.1 | - | 2 | 1447 | |||
| Gene Symbol: 4632433K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BB430846.2 | - | 278 | |||

Zfp784
Genomic Map