siRNA Details
| Gene Symbol: Zfp784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039532.2 | NP_001034621.1 | 2 | 2098 | ||
| GenBank mRNA | AK050452.1 | - | 2 | 2093 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Zfp784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039532.2 | NP_001034621.1 | 2 | 2098 | ||
| GenBank mRNA | AK050452.1 | - | 2 | 2093 | ||
Zfp784
Genomic Map