siRNA Details
| Gene Symbol: Gcnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039560.3 | NP_001034649.1 | 2 | 780 | ||
| GenBank mRNA | AK039414.1 | - | 2 | 782 | ||
| AK132647.1 | - | 1 | 453 | |||
| BC139073.1 | - | 2 | 740 | 8862095 | ||
| BC139076.1 | - | 2 | 740 | 8862098 | ||

Gcnt7
Genomic Map