siRNA Details
| Gene Symbol: Marc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290273.1 | NP_001277202.1 | 7 | 1598 | ||
| GenBank mRNA | AK004989.1 | - | 7 | 1496 | ||
| AK011722.1 | - | 7 | 1479 | |||
| BC028441.1 | - | 5 | 947 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Marc1| Gene Symbol: Marc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290273.1 | NP_001277202.1 | 7 | 1598 | ||
| GenBank mRNA | AK004989.1 | - | 7 | 1496 | ||
| AK011722.1 | - | 7 | 1479 | |||
| BC028441.1 | - | 5 | 947 | |||