siRNA Details
| Gene Symbol: 1110008L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025373.1 | NP_079649.1 | 1 | 1235 | ||
| GenBank mRNA | AK031144.1 | - | 1 | 613 | ||
| AK045209.1 | - | 1 | 1123 | |||
| AK045434.1 | - | 1 | 603 | |||
| AK151976.1 | - | 1 | 1235 | |||
| AK152028.1 | - | 1 | 1235 | |||
| BC034876.1 | - | 1 | 87 | 1512335 | ||

1110008L16Rik
Genomic Map