siRNA Details
| Gene Symbol: Bola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175103.3 | NP_780312.1 | 2 | 118 | ||
| GenBank mRNA | AK002662.1 | - | 2 | 121 | ||
| AK003922.1 | - | 2 | 121 | |||
| AK028165.1 | - | 2 | 132 | |||
| AK140279.1 | - | 199 | ||||
| BC045523.1 | - | 2 | 136 | 6397886 | ||
| BC061197.1 | - | 2 | 146 | 6707992 | ||

Bola2
Genomic Map