siRNA Details
| Gene Symbol: Commd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033132.3 | NP_001028304.1 | 4 | 653 | ||
| NM_001168592.1 | NP_001162064.1 | 3 | 641 | |||
| GenBank mRNA | AK004316.1 | - | 4 | 627 | ||
| AK020234.1 | - | 4 | 626 | |||
| AK045407.1 | - | 3128 | ||||
| CK128085.1 | - | 1 | 404 | |||
| CK621880.1 | - | 3 | 604 | |||

Commd6
Genomic Map