siRNA Details
| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076681.2 | NP_001070149.1 | 4 | 500 | ||
| GenBank mRNA | AK144380.1 | - | 4 | 501 | ||
| BC130224.1 | - | 4 | 491 | 40141726 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyrm9| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076681.2 | NP_001070149.1 | 4 | 500 | ||
| GenBank mRNA | AK144380.1 | - | 4 | 501 | ||
| BC130224.1 | - | 4 | 491 | 40141726 | ||