siRNA Details
| Gene Symbol: Inafm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136270.1 | NP_001129742.1 | 1 | 112 | ||
| GenBank mRNA | CX734215.1 | - | 1 | 142 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inafm1| Gene Symbol: Inafm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136270.1 | NP_001129742.1 | 1 | 112 | ||
| GenBank mRNA | CX734215.1 | - | 1 | 142 | ||