siRNA Details
| Gene Symbol: 1700001K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025488.2 | NP_079764.2 | 5 | 822 | ||
| GenBank mRNA | AK005606.1 | - | 5 | 815 | ||
| AK006358.1 | - | 5 | 818 | |||
| BC048462.1 | - | 5 | 803 | 6774571 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001K19Rik| Gene Symbol: 1700001K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025488.2 | NP_079764.2 | 5 | 822 | ||
| GenBank mRNA | AK005606.1 | - | 5 | 815 | ||
| AK006358.1 | - | 5 | 818 | |||
| BC048462.1 | - | 5 | 803 | 6774571 | ||