siRNA Details
Gene Symbol: 1700001K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025488.2 | NP_079764.2 | 3 | 124 | ||
GenBank mRNA | AK005606.1 | - | 3 | 114 | ||
AK006098.1 | - | 3 | 125 | |||
AK006358.1 | - | 3 | 118 | |||
BC048462.1 | - | 3 | 105 | 6774571 |
