siRNA Details
| Gene Symbol: Klk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019175.1 | NP_062048.1 | 7 | 876 | ||
| XM_006228982.2 | XP_006229044.1 | 7 | 983 | |||
| GenBank mRNA | AF016269.1 | - | 7 | 876 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk6| Gene Symbol: Klk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019175.1 | NP_062048.1 | 7 | 876 | ||
| XM_006228982.2 | XP_006229044.1 | 7 | 983 | |||
| GenBank mRNA | AF016269.1 | - | 7 | 876 | ||