siRNA Details
| Gene Symbol: Gm7616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101600.1 | NP_001095070.1 | 3 | 406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7616| Gene Symbol: Gm7616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101600.1 | NP_001095070.1 | 3 | 406 | ||