siRNA Details
Gene Symbol: Rpl31-ps12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258458.1 | NP_001245387.1 | 1 | 134 | ||
GenBank mRNA | BY609979.1 | - | 1 | 56 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rpl31-ps12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258458.1 | NP_001245387.1 | 1 | 134 | ||
GenBank mRNA | BY609979.1 | - | 1 | 56 |