siRNA Details
| Gene Symbol: Gm7694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198955.1 | NP_001185884.1 | 3 | 3805 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7694| Gene Symbol: Gm7694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198955.1 | NP_001185884.1 | 3 | 3805 | ||