siRNA Details
| Gene Symbol: Gm7714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110779.1 | NP_001104249.1 | 3 | 412 | ||
| GenBank mRNA | AK137735.1 | - | 3 | 414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7714| Gene Symbol: Gm7714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110779.1 | NP_001104249.1 | 3 | 412 | ||
| GenBank mRNA | AK137735.1 | - | 3 | 414 | ||