siRNA Details
| Gene Symbol: Esf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081090.1 | NP_001074559.1 | 4 | 1220 | ||
| GenBank mRNA | AK132230.1 | - | 3 | 1142 | ||
| BC010672.1 | - | 4 | 1191 | |||
| BC021330.1 | - | 4 | 1239 | |||
| BC038028.1 | - | 4 | 562 | |||
| BC055464.1 | - | 3 | 1125 | |||
| BC096029.1 | - | 4 | 1217 | |||
| BC150893.1 | - | 4 | 1186 | 9087804 | ||
| BC157979.1 | - | 4 | 1186 | 9007299 | ||

Esf1
Genomic Map