siRNA Details
| Gene Symbol: Zscan25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081431.1 | NP_001074900.1 | 4 | 826 | ||
| GenBank mRNA | BC150878.1 | - | 4 | 817 | 9087789 | |
| BC151110.1 | - | 4 | 817 | 9088023 | ||
| BC172035.1 | - | 4 | 821 | |||

Zscan25
Genomic Map