siRNA Details
| Gene Symbol: Sepp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083911.1 | NP_001077380.1 | 3 | 431 | ||
| NM_019192.2 | NP_062065.2 | 3 | 359 | |||
| GenBank mRNA | BC059137.1 | - | 3 | 360 | ||
| BC072539.1 | - | 3 | 348 | 7104195 | ||
| D25221.1 | - | 3 | 340 | |||
| DN933732.1 | - | 3 | 679 | |||
| M63574.1 | - | 319 | ||||

Sepp1
Genomic Map