siRNA Details
Gene Symbol: 4933415F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025746.2 | NP_080022.2 | 1 | 1001 | ||
GenBank mRNA | AK005938.1 | - | 1 | 528 | ||
AK016819.1 | - | 1 | 1002 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933415F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025746.2 | NP_080022.2 | 1 | 1001 | ||
GenBank mRNA | AK005938.1 | - | 1 | 528 | ||
AK016819.1 | - | 1 | 1002 |