siRNA Details
| Gene Symbol: 4933415F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025746.2 | NP_080022.2 | 1 | 1748 | ||
| GenBank mRNA | AK005938.1 | - | 1 | 1275 | ||
| AV040646.2 | - | 1 | 257 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933415F23Rik| Gene Symbol: 4933415F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025746.2 | NP_080022.2 | 1 | 1748 | ||
| GenBank mRNA | AK005938.1 | - | 1 | 1275 | ||
| AV040646.2 | - | 1 | 257 | |||