siRNA Details
| Gene Symbol: Gm8817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101606.1 | NP_001095076.1 | 4 | 374 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI592229.1 | - | 257 | Chr. - - [-] on Build | ||

Gm8817
Genomic Map