siRNA Details
| Gene Symbol: 9030624G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256489.1 | NP_001243418.1 | 4 | 1227 | ||
| GenBank mRNA | AK018572.1 | - | 4 | 1205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030624G23Rik| Gene Symbol: 9030624G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256489.1 | NP_001243418.1 | 4 | 1227 | ||
| GenBank mRNA | AK018572.1 | - | 4 | 1205 | ||