siRNA Details
| Gene Symbol: Rbm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025776.2 | NP_080052.1 | 11 | 1800 | ||
| GenBank mRNA | AK018452.1 | - | 11 | 1823 | ||
| AK045116.1 | - | 11 | 1821 | |||
| AK150271.1 | - | 11 | 1813 | |||
| AK151758.1 | - | 11 | 1813 | |||
| AK152252.1 | - | 11 | 1813 | |||
| AK166600.1 | - | 11 | 1821 | |||
| AK167509.1 | - | 11 | 1840 | |||
| AK168919.1 | - | 11 | 1801 | |||
| BC042691.1 | - | 11 | 1819 | |||
| BC080205.1 | - | 11 | 1798 | 3709760 | ||

Rbm22
Genomic Map