siRNA Details
| Gene Symbol: Syce1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048145.1 | NP_001041610.1 | 11 | 784 | ||
| NM_001286494.1 | NP_001273423.1 | 10 | 721 | |||
| NM_029139.1 | NP_083415.1 | 12 | 848 | |||
| GenBank mRNA | AK015606.1 | - | 12 | 844 | ||
| AY899935.1 | - | 11 | 784 | |||
| AY899936.1 | - | 12 | 848 | |||
| BC100478.1 | - | 10 | 702 | |||

Syce1l
Genomic Map