siRNA Details
| Gene Symbol: Esp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244651.1 | NP_001231580.1 | 3 | 479 | ||
| GenBank mRNA | AB306993.1 | - | 3 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esp15| Gene Symbol: Esp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244651.1 | NP_001231580.1 | 3 | 479 | ||
| GenBank mRNA | AB306993.1 | - | 3 | 479 | ||